siRNA Details
Gene Symbol: Gm26839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131831.1 | - | 3 | 326 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707656.1 | - | 3 | 322 | Chr. 12 - 99563844 - 99619912 [+] on Build GRCm38 |
