siRNA Details
Gene Symbol: Gm20337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045057.1 | 3 | 1057 | |||
GenBank mRNA | AK145430.1 | - | 3 | 1056 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY756110.1 | - | 3 | 619 | Chr. 12 - 80942673 - 80944916 [-] on Build GRCm38 |
