siRNA Details
Gene Symbol: Gm20337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045057.1 | 1 | 165 | |||
GenBank mRNA | AK145430.1 | - | 1 | 164 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ963819.1 | - | 1 | 140 | Chr. 12 - 80942857 - 80945374 [-] on Build GRCm38 |
