siRNA Details
Gene Symbol: Popdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK171056.1 | - | 1291 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY534803.1 | - | 1 | 82 | Chr. 16 - 38378433 - 38378870 [+] on Build GRCm38 |
