siRNA Details
Gene Symbol: 4933406D12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040502.1 | 4 | 454 | |||
GenBank mRNA | AK016684.1 | - | 4 | 455 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716371.1 | - | 4 | 455 | Chr. 2 - 146542931 - 146546108 [+] on Build GRCm38 |
