siRNA Details
Gene Symbol: 9430014N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045737.1 | 2 | 256 | |||
GenBank mRNA | AK020418.1 | - | 2 | 257 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721595.1 | - | 2 | 257 | Chr. 15 - 93905965 - 93927111 [-] on Build GRCm38 |
