siRNA Details
Gene Symbol: 9430014N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045737.1 | 3, 4 | 885 | |||
GenBank mRNA | AK020418.1 | - | 3, 4 | 886 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9430014N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045737.1 | 3, 4 | 885 | |||
GenBank mRNA | AK020418.1 | - | 3, 4 | 886 |