siRNA Details
Gene Symbol: 9430012M22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020412.1 | - | 1 | 226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721588.1 | - | 1 | 226 | Chr. 3 - 55290944 - 55291595 [+] on Build GRCm38 |
