siRNA Details
Gene Symbol: 1700021P04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006227.1 | - | 2 | 297 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706313.1 | - | 2 | 297 | Chr. 19 - 24064793 - 24066452 [+] on Build GRCm38 |
