siRNA Details
Gene Symbol: 1700021N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006223.1 | - | 1 | 180 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706309.1 | - | 1 | 180 | Chr. 19 - 48240860 - 48241114 [-] on Build GRCm38 |
