siRNA Details
Gene Symbol: 6530403G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018318.1 | - | 1 | 1259 | ||
AK045988.1 | - | 1 | 1255 | |||
AK084809.1 | - | 1 | 1265 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB750740.1 | - | 1 | 246 | Chr. 10 - 7791388 - 7791807 [-] on Build GRCm38 |
