siRNA Details
Gene Symbol: Reln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK082390.1 | - | 1 | 2815 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB647157.1 | - | 1 | 323 | Chr. 5 - 21890934 - 21891404 [-] on Build GRCm38 |
