siRNA Details
Gene Symbol: Gm20356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040450.1 | 4 | 493 | |||
GenBank mRNA | AK139930.1 | - | 4 | 494 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY726125.1 | - | 4 | 494 | Chr. 3 - 74011032 - 74017277 [-] on Build GRCm38 |
