siRNA Details
Gene Symbol: 2200007N16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019048.1 | - | 1 | 161 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708532.1 | - | 1 | 161 | Chr. 7 - 24666733 - 24666986 [+] on Build GRCm38 |
