siRNA Details
Gene Symbol: Zfp866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133399.1 | - | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617032.1 | - | 2 | 52 | Chr. 8 - 69763368 - 69791175 [-] on Build GRCm38 |
