siRNA Details
Gene Symbol: 2900002H24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019300.1 | - | 1 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712438.1 | - | 1 | 152 | Chr. 1 - 38322228 - 38322499 [-] on Build GRCm38 |
