siRNA Details
Gene Symbol: 1700063I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018872.1 | - | 1 | 597 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU939509.1 | - | 1 | 592 | Chr. 1 - 168675272 - 168675877 [-] on Build GRCm38 |
