siRNA Details
Gene Symbol: 4930517J16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015815.1 | - | 1 | 284 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715357.1 | - | 1 | 281 | Chr. 4 - 44669333 - 44670050 [+] on Build GRCm38 |
