siRNA Details
Gene Symbol: 2210416O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045499.1 | 1 | 13 | |||
GenBank mRNA | AK008958.1 | - | 1 | 14 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708845.1 | - | 1 | 14 | Chr. 11 - 88098058 - 88099419 [+] on Build GRCm38 |
