siRNA Details
Gene Symbol: 4930517M08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015816.1 | - | 3 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715360.1 | - | 3 | 412 | Chr. 17 - 4634978 - 4637455 [+] on Build GRCm38 |
