siRNA Details
Gene Symbol: 1700029J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040494.1 | 1 | 170 | |||
GenBank mRNA | AK006500.1 | - | 1 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706535.1 | - | 1 | 171 | Chr. 16 - 93396918 - 93458872 [-] on Build GRCm38 |
