siRNA Details
Gene Symbol: Slc25a51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK172225.1 | - | 1 | 525 | ||
BC026435.1 | - | 1 | 21 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE533311.1 | - | 1 | 421 | Chr. 4 - 45408124 - 45408651 [-] on Build GRCm38 |
