siRNA Details
Gene Symbol: C78283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144862.1 | - | 2 | 544 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734523.1 | - | 2 | 542 | Chr. 5 - 73946383 - 73948259 [+] on Build GRCm38 |
