siRNA Details
Gene Symbol: Gm38483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053109.1 | - | 1488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB529520.2 | - | 3 | 550 | Chr. 11 - 100690772 - 100697520 [+] on Build GRCm38 |
