siRNA Details
Gene Symbol: A730080H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043280.1 | - | 1 | 226 | ||
AK047900.1 | - | 1 | 1269 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU513811.1 | - | 379 | Chr. - - [-] on Build |
