siRNA Details
Gene Symbol: 5830426K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017949.1 | - | 1 | 271 | ||
AK041807.1 | - | 1 | 1313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717837.1 | - | 1 | 271 | Chr. 11 - 87341989 - 87342809 [-] on Build GRCm38 |
