siRNA Details
Gene Symbol: 1700011B04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045616.1 | 3 | 376 | |||
GenBank mRNA | AI606400.1 | - | 2 | 303 | ||
AK005856.1 | - | 3 | 378 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705998.1 | - | 3 | 377 | Chr. 13 - 35181623 - 35188009 [+] on Build GRCm38 |
