siRNA Details
Gene Symbol: 5830428M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038060.1 | 4 | 258 | |||
NR_038061.1 | 4 | 252 | ||||
GenBank mRNA | AI060956.1 | - | 4 | 233 | ||
AK039829.1 | - | 4 | 255 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB634306.1 | - | 4 | 255 | Chr. 12 - 69465168 - 69475547 [-] on Build GRCm38 |
