siRNA Details
Gene Symbol: 6030451C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020069.1 | - | 1 | 163 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718136.1 | - | 1 | 158 | Chr. 4 - 105200739 - 105201413 [+] on Build GRCm38 |
