siRNA Details
Gene Symbol: 4930557J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040703.1 | 1 | 229 | |||
GenBank mRNA | AK016164.1 | - | 1 | 231 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715767.1 | - | 231 | Chr. - - [-] on Build |
