siRNA Details
Gene Symbol: A630095N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080399.1 | - | 3 | 503 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639432.1 | - | 3 | 503 | Chr. 1 - 75220131 - 75223654 [-] on Build GRCm38 |
