siRNA Details
Gene Symbol: 4933406G16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046037.1 | 3 | 1017 | |||
GenBank mRNA | AK016692.1 | - | 3 | 1018 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB273454.1 | - | 3 | 483 | Chr. 11 - 19065810 - 19068583 [+] on Build GRCm38 |
