siRNA Details
Gene Symbol: 9530049O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020601.1 | - | 1 | 571 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722137.1 | - | 1 | 571 | Chr. 5 - 31957067 - 31957769 [+] on Build GRCm38 |
