siRNA Details
Gene Symbol: 9530050K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020603.1 | - | 1 | 120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722142.1 | - | 1 | 117 | Chr. 12 - 100079320 - 100079760 [-] on Build GRCm38 |
