siRNA Details
Gene Symbol: 1700006F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045621.1 | 2, 3 | 164 | |||
GenBank mRNA | AK005671.1 | - | 2, 3 | 166 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705857.1 | - | 2, 3 | 166 | Chr. 14 - 119749225 - 119751564 [+] on Build GRCm38 |
