siRNA Details
Gene Symbol: 4930519H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045974.1 | 5, 6 | 838 | |||
GenBank mRNA | AK076692.1 | - | 5, 6 | 839 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV260727.2 | - | 4, 5 | 505 | Chr. 5 - 15863928 - 15875171 [-] on Build GRCm38 |
