siRNA Details
Gene Symbol: Gm4668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041339.1 | - | 4 | 902 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB227220.2 | - | 3 | 659 | Chr. 9 - 116190822 - 116194870 [+] on Build GRCm38 |
