siRNA Details
Gene Symbol: Gm13982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040062.1 | - | 2 | 136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB635101.1 | - | 2 | 133 | Chr. 2 - 117414515 - 117417695 [-] on Build GRCm38 |
