siRNA Details
Gene Symbol: Gm16096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089373.1 | - | 804 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY764361.1 | - | 3 | 698 | Chr. 9 - 40780740 - 40783425 [-] on Build GRCm38 |
