siRNA Details
Gene Symbol: 4831407H17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019510.1 | - | 4 | 447 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714113.1 | - | 4 | 447 | Chr. 14 - 30458036 - 30462765 [+] on Build GRCm38 |
