siRNA Details
Gene Symbol: Ascc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011671.1 | - | 1 | 262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ944505.1 | - | 1 | 200 | Chr. 11 - 4683962 - 4684827 [+] on Build GRCm38 |
