siRNA Details
Gene Symbol: 4930421J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015171.1 | - | 1 | 342 | ||
AK076722.1 | - | 1 | 343 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV261392.2 | - | 1 | 238 | Chr. 12 - 110737262 - 110738251 [-] on Build GRCm38 |
