siRNA Details
Gene Symbol: 4930421P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015174.1 | - | 1 | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714724.1 | - | 1 | 391 | Chr. 9 - 31028972 - 31029832 [-] on Build GRCm38 |
