siRNA Details
Gene Symbol: Gm6277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045421.1 | 4 | 434 | |||
GenBank mRNA | AK140316.1 | - | 4 | 435 | ||
BC030050.1 | - | 4 | 394 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX529752.1 | - | 4 | 395 | Chr. 18 - 11821953 - 11839337 [-] on Build GRCm38 |
