siRNA Details
Gene Symbol: Gm12602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144841.1 | - | 2 | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734487.1 | - | 2 | 173 | Chr. 4 - 89100664 - 89128226 [-] on Build GRCm38 |
