siRNA Details
Gene Symbol: Gm10067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147851.1 | - | 5 | 595 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY741011.1 | - | 5 | 595 | Chr. 8 - 9878348 - 9919463 [-] on Build GRCm38 |
