siRNA Details
Gene Symbol: 5830416I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045384.1 | 3 | 2562 | |||
GenBank mRNA | AK080054.1 | - | 1 | 954 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB219247.2 | - | 1 | 568 | Chr. 5 - 64049590 - 64050228 [+] on Build GRCm38 |
