siRNA Details
Gene Symbol: 5830449F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018000.1 | - | 1 | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717925.1 | - | 1 | 329 | Chr. 16 - 23280161 - 23280685 [+] on Build GRCm38 |
