siRNA Details
Gene Symbol: Gm39168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078582.1 | - | 2, 3 | 303 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB623744.1 | - | 2, 3 | 303 | Chr. 8 - 46299131 - 46324927 [+] on Build GRCm38 |
