siRNA Details
Gene Symbol: 4930557B06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016157.1 | - | 1 | 125 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715760.1 | - | 1 | 126 | Chr. 5 - 113142455 - 113142822 [+] on Build GRCm38 |
